missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POF1B Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93152-0.1ml
This item is not returnable.
View return policy
Description
POF1B Polyclonal antibody specifically detects POF1B in Human, Rat samples. It is validated for Western Blot
Specifications
| POF1B | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| FLJ22792, POF2B, premature ovarian failure, 1B, protein POF1B | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 330-560 of human POF1B (NP_079197.3). LVLSTFSNIREELGHLQNDMTSLENDKMRLEKDLSFKDTQLKEYEELLASVRANNHQQQQGLQDSSSKCQALEENNLSLRHTLSDMEYRLKELEYCKRNLEQENQNLRMQVSETCTGPMLQAKMDEIGNHYTEMVKNLRMEKDREICRLRSQLNQYHKDVSKREGSCSDFQFKLHELTSLLEEKDSLIKRQSEELSKLRQEIYSSHNQPSTGGRTTITTKKYRTQYPILGL | |
| 0.1 mL | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 79983 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion