missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ POF1B Polyclonal Antibody

Product Code. 15799620 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15799620 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15799620 Supplier Invitrogen™ Supplier No. PA551671

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: VSETCTGPML QAKMDEIGNH YTEMVKNLRM EKDREICRLR SQLNQYHKDV SKREGSCSDF QFKLHELTSL LEEKDSLIKR QSEELSKLRQ EIYSSHNQPS TGGRTTITTK KYRTQYPILG LLYDDYEYIP PGSETQTI Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.

Plays a key role in the organization of epithelial monolayers by regulating the actin cytoskeleton. May be involved in ovary development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POF1B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene POF1B
Gene Accession No. Q8WVV4
Gene Alias 2310066B14Rik; POF; POF1B; POF1B actin binding protein; POF1B, actin binding protein; POF2B; premature ovarian failure 1B; premature ovarian failure protein 1B; premature ovarian failure, 1B; protein POF1B; RGD1560798
Gene Symbols POF1B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human POF1B. Recombinant protein control fragment (Product #RP-92126).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79983
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.