missing translation for 'onlineSavingsMsg'
Learn More

POFUT2 Antibody (OTI3A12), Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18446895 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18446895 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18446895 Supplier Novus Biologicals Supplier No. NBP273512AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

POFUT2 Monoclonal antibody specifically detects POFUT2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen POFUT2
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone OTI3A12
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias C21orf80, chromosome 21 open reading frame 80, FUT13EC 2.4.1.221, GDP-fucose protein O-fucosyltransferase 2, KIAA0958, O-FucT-2, Peptide-O-fucosyltransferase 2, protein O-fucosyltransferase 2
Host Species Mouse
Immunogen Full length recombinant protein of human POFUT2 (NP_598368) produced in HEK293T cells.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 23275
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.