missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POFUT2 Antibody (OTI3A12), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-73512CL1
This item is not returnable.
View return policy
Description
POFUT2 Monoclonal antibody specifically detects POFUT2 in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Western Blot,CyTOF
Specifications
| POFUT2 | |
| Monoclonal | |
| CoraFluor 1 | |
| C21orf80, chromosome 21 open reading frame 80, FUT13EC 2.4.1.221, GDP-fucose protein O-fucosyltransferase 2, KIAA0958, O-FucT-2, Peptide-O-fucosyltransferase 2, protein O-fucosyltransferase 2 | |
| Full length human recombinant protein of human POFUT2(NP_598368) produced in HEK293T cell. | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 23275 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2a |
| Flow Cytometry, Western Blot, CyTOF | |
| OTI3A12 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction