missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POGZ Antibody (PCRP-POGZ-1B2), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-24195MFV500
This item is not returnable.
View return policy
Description
POGZ Monoclonal antibody specifically detects POGZ in Human samples. It is validated for Flow Cytometry
Specifications
| POGZ | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| KIAA0461ZNF635, pogo transposable element with ZNF domain, putative protein product of Nbla00003, SUHW5MGC71543, Suppressor of hairy wing homolog 5, Zinc finger protein 280E, Zinc finger protein 635, ZNF280EZNF635m | |
| Recombinant full-length human POGZ protein | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| PCRP-POGZ-1B2 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 23126 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction