missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polaprezinc, MedChemExpress
Description
Polaprezinc is an orally bioavailable chelate composed of zinc and L-carnosine, with potential gastroprotective, anti-oxidant, anti-ulcer and anti-inflammatory activities.
Specifications
Specifications
| Chemical Name or Material | Polaprezinc |
| Physical Form | Solid |
| CAS | 107667-60-7 |
| Color | White |
| Quantity | 10 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C9H11N4O3Zn |
| For Use With (Application) | COVID-19-immunoregulation |
| Synonym | Zinc L-carnosine |
| Solubility Information | H2O : 3.37 mg/mL (11.68 mM; ultrasonic and warming and adjust pH to 1-2 with HCl) ∣DMSO : < 1 mg/mL (insoluble or slightly soluble) |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?