missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ PolarScreen™ Vitamin D Receptor Competitor Assay Kit, Red

Product Code. 15677858
Change view
Click to view available options
Quantity:
800 x 20 μL assays
Unit Size:
Pack of 800
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15677858 800 x 20 μL assays Pack of 800
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15677858 Supplier Thermo Scientific™ Supplier No. A15907

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The PolarScreen™ Vitamin D Receptor Competitor Assay Kit, Red, provides a sensitive and efficient method for high-throughput, fluorescence polarization-based screening of potential vitamin D receptor (VDR) ligands.

The PolarScreen™ Vitamin D Receptor Competitor Assay Kit, Red, provides a sensitive and efficient method for high-throughput, fluorescence polarization-based screening of potential vitamin D receptor (VDR) ligands. The kit uses insect cell-expressed, human full-length VDR (also available separately) and a novel, high-affinity, fluorescent VDR ligand (Fluormone™ VDR Red) in a homogenous, mix-and-read assay format.

This kit produces a red color read-out and is suitable to determine the IC50s of competitive compounds. For additional information or assistance contact us at drugdiscoverytech@lifetech.com.

The PolarScreen Vitamine D Receptor Competitor Assay is:

  • Complete & ready-to-use out of the box—all reagents included, just add your test compounds
  • High throughput compatible—fluorescence polarization assays in a 384-well plate
  • Convenient—components available in bulk sizes for larger screening needs

How the Assay Works

VDR is added to a fluorescent VDR ligand (Fluormone VDR Red) to form a receptor/tracer complex, resulting in a high polarization value. This complex is then added to individual test compounds in microwell plates. Competitors displace the tracer from the complex, causing the fluorescent ligand to tumble more rapidly during its fluorescence lifetime, resulting in a low polarization value. Noncompetitors will not displace the fluorescent ligand from the complex; therefore, the polarization value will remain high. The shift in polarization value in the presence of test compounds is used to determine relative affinity of test compounds for VDR (see figure).

Contact us for bulk quantities or for assistance with other nuclear receptor assay needs, from assay development to library screening and beyond.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Competitive Binding Assay, Nuclear Receptor Assay
Quantity 800 x 20 μL assays
Content And Storage • VDR Recombinant Human Protein, 1 tube (store at -68°C to -85°C)
• Fluormone VDR Red Tracer, 1 tube (-5°C to -30°C)
• VDR Red Screening Buffer, 3 bottles (-5°C to -30°C)
• DTT, 1 tube (-5°C to -30°C)
Product Line PolarScreen
Ligand VDR
Product Type Vitamin D Receptor Competitor Assay Kit
No. of Assays 800 x 20 μL Assays

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.