missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POLD3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21935-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes the 66-kDa subunit of DNA polymerase delta. DNA polymerase delta possesses both polymerase and 3′ to 5′ exonuclease activity and plays a critical role in DNA replication and repair. The encoded protein plays a role in regulating the activity of DNA polymerase delta through interactions with other subunits and the processivity cofactor proliferating cell nuclear antigen (PCNA). Alternatively spliced transcript variants have been observed for this gene.Specifications
| POLD3 | |
| Polyclonal | |
| Unconjugated | |
| POLD3 | |
| DNA polymerase delta subunit 3, KIAA0039, P66, P68, POLD3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10714, 67967 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunofluorescence, Immunocytochemistry | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q15054, Q9EQ28 | |
| POLD3 | |
| POLD3 Fusion Protein Ag16570 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction