missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POLDIP2 Antibody (PSH01-20), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32839
This item is not returnable.
View return policy
Description
POLDIP2 Monoclonal antibody specifically detects POLDIP2 in Human,Rat samples. It is validated for Western Blot
Specifications
| POLDIP2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000 | |
| DKFZP586F1524, p38, PDIP38DKFZp586F1524,38 kDa DNA polymerase delta interaction protein, POLD4, polymerase (DNA-directed), delta interacting protein 2, polymerase delta interacting protein 38, polymerase delta-interacting protein 2 | |
| Recombinant protein within human POLDIP2 aa 31-368/ 368. (Uniprot: Q9Y2S7) | |
| 100 μL | |
| Endocrinology | |
| 26073 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| PSH01-20 | |
| Unconjugated | |
| PBS (pH7.4), 0.1% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction