missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ POLDIP2 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142650
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:8,000.
POLDIP2 is a protein that interacts with the DNA polymerase delta p50 subunit, as well as with proliferating cell nuclear antigen. The encoded protein maybe play a role in the ability of the replication fork to bypass DNA lesions. Alternative splicing results in multiple transcript variants.
Specifications
| POLDIP2 | |
| Polyclonal | |
| Unconjugated | |
| Poldip2 | |
| 1300003F06Rik; 38 kDa DNA polymerase delta interaction protein; DKFZp586F1524; DNA polymerase delta interacting protein 2; DNA-directed polymerase delta interacting protein 2; HSPC017; mitogenin 1; mitogenin I; p38; PDIP38; POLD4; POLDIP2; polymerase (DNA) delta interacting protein 2; polymerase (DNA-directed), delta interacting protein 2; polymerase delta interacting protein 38; polymerase delta-interacting protein 2 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 26073, 287544, 67811 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q91VA6, Q9Y2S7 | |
| Poldip2 | |
| Peptide with sequence KTHTYYQVLIDARDC. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction