missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ POLE Recombinant Rabbit Monoclonal Antibody (HL2150)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA550403

Product Code. 18016441

  • € 491.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

POLE Recombinant Monoclonal Antibody for Western Blot, IHC (P)

POLE (Polepsi) is present in proliferating cells and is implicated in repair of UV-damaged DNA.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

POLE
Recombinant Monoclonal
2 mg/mL
PBS with no preservative
Q07864
POLE
Recombinant fragment of human POLE.
100 μL
Primary
Human
Antibody
IgG
Immunohistochemistry (Paraffin), Western Blot
HL2150
Unconjugated
POLE
CRCS12; DNA polymerase epsilon catalytic subunit A; DNA polymerase epsilon catalytic subunit protein; DNA polymerase epsilon, catalytic subunit; DNA polymerase II subunit A; DNA-directed DNA polymerase epsilon; FILS; Pole; POLE1; pol-epsilon; polymerase (DNA directed), epsilon; polymerase (DNA directed), epsilon, catalytic subunit; polymerase (DNA) epsilon, catalytic subunit
Rabbit
Protein A
RUO
5426
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.