missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POLE4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93562-0.1ml
This item is not returnable.
View return policy
Description
POLE4 Polyclonal antibody specifically detects POLE4 in Human samples. It is validated for Western Blot
Specifications
| POLE4 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| DNA polymerase epsilon p12 subunit, DNA polymerase epsilon subunit 4, DNA polymerase epsilon subunit p12, DNA polymerase II subunit 4, EC 2.7.7.7, p12, polymerase (DNA-directed), epsilon 4 (p12 subunit), YHHQ1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-117 of human POLE4 (NP_063949.2). MAAAAAAGSGTPREEEGPAGEAAASQPQAPTSVPGARLSRLPLARVKALVKADPDVTLAGQEAIFILARAAELFVETIAKDAYCCAQQGKRKTLQRRDLDNAIEAVDEFAFLEGTLD | |
| 0.1 mL | |
| DNA Polymerases, DNA Repair | |
| 56655 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction