Learn More
POLI Mouse anti-Human, Clone: OTI4C12, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF801490
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Error-prone DNA polymerase specifically involved in DNA repair. Plays an important role in translesion synthesis, where the normal high-fidelity DNA polymerases cannot proceed and DNA synthesis stalls. Favors Hoogsteen base-pairing in the active site. Inserts the correct base with high-fidelity opposite an adenosine template. Exhibits low fidelity and efficiency opposite a thymidine template, where it will preferentially insert guanosine. May play a role in hypermutation of immunogobulin genes. Forms a Schiff base with 5′-deoxyribose phosphate at abasic sites, but may not have lyase activity.Specifications
| POLI | |
| Monoclonal | |
| Unconjugated | |
| POLI | |
| RAD30B, RAD3OB, eta2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 11201 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI4C12 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q9UNA4 | |
| POLI | |
| Human recombit protein fragment corresponding to amino acids 458-740 of human POLI produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.