missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POLI Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13635-1-AP-20UL
This item is not returnable.
View return policy
Description
This is a paternally expressed imprinted gene that encodes transcripts containing two overlapping open reading frames start codon, have been reported for this gene. Increased expression of this gene is associated with hepatocellular carcinomas.Specifications
| POLI | |
| Polyclonal | |
| Unconjugated | |
| POLI | |
| DNA polymerase iota, Eta2, POLI, polymerase (DNA directed) iota, RAD30 homolog B, RAD30B, RAD3OB | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11201, 26447, 291526 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot | |
| 0.18 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6R3M4, Q9UNA4 | |
| Poli | |
| POLI Fusion Protein Ag4563 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction