missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AP2 alpha Antibody (AP2a 8G8/5), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2639FR
This item is not returnable.
View return policy
Description
AP2 alpha Monoclonal specifically detects AP2 alpha in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| AP2 alpha | |
| Monoclonal | |
| DyLight 680 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| TFAP2A | |
| Bacterialy expressed AP-2a/AP-2g fusion protein. | |
| 0.1 mL | |
| Cancer, Transcription Factors and Regulators | |
| 7020 | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| AP2a 8G8/5 | |
| Western Blot, Immunohistochemistry | |
| Activating enhancer-binding protein 2-alpha, Activator protein 2, AP-2 transcription factor, AP2-alpha, AP2TFAP-2alpha, AP-2transcription factor AP-2 alpha (activating enhancer-binding protein 2 alpha), BOFS, FLJ51761, TFAP2, transcription factor AP-2 alpha (activating enhancer binding protein 2 alpha), transcription factor AP-2-alpha | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| AP2a 8G8/5 is specific for the alpha form of AP-2.Antigen recovery using microwave or pressure cooking required for paraffin embedded tissue. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction