Learn More
POLR2D Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97517MFV500
This item is not returnable.
View return policy
Description
POLR2D Polyclonal antibody specifically detects POLR2D in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| Western Blot, Immunoprecipitation | |
| Western Blot, Immunoprecipitation | |
| 0.1 mL | |
| Stem Cell Markers | |
| 5433 | |
| Purified |
| mFluor Violet 500 SE | |
| Produced in rabbits immunized with E. coli-derived Human POLR2D fragment. | |
| RUO | |
| Primary | |
| Store at 4C in the dark. |