Learn More
POLR2D Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-97517MFV500
Product Code. 18575631
  • € 428.00 / 0.10mL
Description
POLR2D Polyclonal antibody specifically detects POLR2D in Human samples. It is validated for Western Blot, Immunoprecipitation
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Western Blot, Immunoprecipitation
Western Blot, Immunoprecipitation
0.1 mL
Stem Cell Markers
5433
Purified
mFluor Violet 500 SE
Produced in rabbits immunized with E. coli-derived Human POLR2D fragment.
RUO
Primary
Store at 4C in the dark.