missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POLR2D Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97517PE
This item is not returnable.
View return policy
Description
POLR2D Polyclonal antibody specifically detects POLR2D in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| POLR2D | |
| Polyclonal | |
| Western Blot, Immunoprecipitation | |
| DNA-directed RNA polymerase II 16 kDa polypeptide, DNA-directed RNA polymerase II subunit D, DNA-directed RNA polymerase II subunit RPB4, HSRBP4, HSRPB4, polymerase (RNA) II (DNA directed) polypeptide D, RBP4, RNA polymerase II 16 kDa subunit, RNA polymerase II subunit B4, RNA polymerase II subunit hsRBP4, RPB16 | |
| Produced in rabbits immunized with E. coli-derived Human POLR2D fragment. | |
| 0.1 mL | |
| Stem Cell Markers | |
| 5433 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| PE | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction