missing translation for 'onlineSavingsMsg'
Learn More

POLR2F Monoclonal Antibody (OTI2F2), TrueMAB™, OriGene

Product Code. 17538523
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17538523 30 μL 30µL
17518523 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17538523 Supplier Origene Technologies Supplier No. TA807758S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

POLR2F Monoclonal antibody specifically detects POLR2F in Human samples. It is validated for Western Blot

Specifications

Antigen POLR2F
Applications Western Blot
Classification Monoclonal
Clone OTI2F2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene SMCP
Gene Accession No. P49901
Gene Alias 1810060D16Rik; DNA-directed RNA polymerase II subunit F; DNA-directed RNA polymerases I, II, and III 14.4 kDa polypeptide; DNA-directed RNA polymerases I, II, and III subunit RPABC2; HRBP14.4; hsRPB6; Polr2f; POLRF; polymerase (RNA) II (DNA directed) polypeptide F; polymerase (RNA) II subunit F; polymerase II; RNA Pol II; RNA Polymerase II subunit 14.4 kD; RNA polymerase II subunit F; RNA polymerases I, II, and III subunit ABC2; RPABC14.4; RPABC2; RPB14.4; RPB6; RPB6 homolog; RPC15
Host Species Mouse
Immunogen Full length human recombinant protein of human SMCP produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4184
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.