Learn More
POLR2F Mouse, anti-Human, Clone: 2D4D12, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 68511-1-IG-20UL
 View more versions of this product
Product Code. 18025205
  • € 145.00 / 20µL
Description
The POLR2F antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human POLR2F. This antibody recognizes human, mouse antigen. The POLR2F antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
POLR2F
2D4D12
HRBP14.4, POLR2F, POLRF, RPABC14.4, RPABC2, RPB14.4, RPB6, RPB6 homolog, RPC15
20 μL
5435
Store at -20°C. Stable for one year after shipment.
Liquid
Western Blot, ELISA
1000 μg/mL
Mouse
Primary
Human, Mouse
150 kDa