missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POLR2G Polyclonal Antibody

POLR2G Polyclonal Antibody
Brand: Invitrogen PA576201
This item is not returnable.
View return policy
Description
POLR2G Polyclonal Antibody for Western Blot
Specifications
| POLR2G | |
| Polyclonal | |
| Unconjugated | |
| POLR2G | |
| 2410046K11Rik; A230108L04Rik; C76415; DNA directed RNA polymerase II 19 kda polypeptide; DNA-directed RNA polymerase II 19 kDa polypeptide; DNA-directed RNA polymerase II subunit G; DNA-directed RNA polymerase II subunit RPB7; h hsRPB7; hRPB19; hs; hsRPB7; MGC138367; MGC138369; POLR2G; polymerase (RNA) II (DNA directed) polypeptide G; polymerase (RNA) II subunit G; RBP7; RNA polymerase II 19 kDa subunit; RNA polymerase II seventh subunit; RNA polymerase II subunit B7; RNA polymerase II subunit G; RPB19; RPB7; Rpo2-7l | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 117017, 5436, 67710 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P62487, P62488, P62489 | |
| POLR2G | |
| Recombinant fusion protein of human POLR2G. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction