missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POLR2J Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 198.00 - € 468.00
Specifications
| Antigen | POLR2J |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
POLR2J Polyclonal antibody specifically detects POLR2J in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| POLR2J | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Neurodegeneration, Neuroscience | |
| PBS (pH 7.3), 50% glycerol | |
| 5439 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| DNA-directed RNA polymerase II subunit J-1, DNA-directed RNA polymerase II subunit RPB11-a, hRPB14, MGC71910, POLR2J1RNA polymerase II subunit B11-a, polymerase (RNA) II (DNA directed) polypeptide J (13.3kD), polymerase (RNA) II (DNA directed) polypeptide J, 13.3kDa, RNA polymerase II 13.3 kDa subunit, RPB11, RPB11A, RPB11m | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-117 of human POLR2J (NP_006225.1). MNAPPAFESFLLFEGEKKITINKDTKVPNACLFTINKEDHTLGNIIKSQLLKDPQVLFAGYKVPHPLEHKIIIRVQTTPDYSPQEAFTNAITDLISELSLLEERFRVAIKDKQEGIE | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title