Learn More
POLR2J Mouse anti-Human, Clone: OTI1H4, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF810707
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
DNA-dependent RNA polymerase catalyzes the transcription of DNA into RNA using the four ribonucleoside triphosphates as substrates. Component of RNA polymerase II which synthesizes mRNA precursors and many functional non-coding RNAs. Pol II is the central component of the basal RNA polymerase II transcription machinery. It is composed of mobile elements that move relative to each other. RPB11 is part of the core element with the central large cleft.Specifications
| POLR2J | |
| Monoclonal | |
| Unconjugated | |
| POLR2J | |
| POLR2J1, RPB11, RPB11A, RPB11m, hRPB14 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 5439 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI1H4 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P52435 | |
| POLR2J | |
| Full length human recombit protein of human POLR2J produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.