missing translation for 'onlineSavingsMsg'
Learn More

POLR2K Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

€ 198.00 - € 468.00

Specifications

Antigen POLR2K
Dilution Western Blot 1:500-1:2000
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18659270
View Documents
Novus Biologicals
NBP2-93400-0.02ml
0.02 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
18699970
View Documents
Novus Biologicals
NBP2-93400-0.1ml
0.1 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

POLR2K Polyclonal antibody specifically detects POLR2K in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications

Specifications

POLR2K
Western Blot
Unconjugated
Rabbit
DNA Repair, DNA replication Transcription Translation and Splicing
PBS (pH 7.3), 50% glycerol
5440
IgG
Affinity purified
Western Blot 1:500-1:2000
Polyclonal
Purified
RUO
Human, Mouse, Rat
ABC10-alpha, DNA directed RNA polymerases I, II, and III 7.0 kda polypeptide, DNA-directed RNA polymerase II subunit K, DNA-directed RNA polymerases I, II, and III subunit RPABC4, hRPB7.0, hsRPB10a, polymerase (RNA) II (DNA directed) polypeptide K, 7.0kDa, RNA polymerase II 7.0 kDa subunit, RNA polymerases I, II, and III subunit ABC4, RPABC4, RPB10alphapolymerase (RNA) II (DNA directed) polypeptide K (7.0kD), RPB12, RPB7.0
Recombinant fusion protein containing a sequence corresponding to amino acids 1-58 of human POLR2K (NP_005025.1). MDTQKDVQPPKQQPMIYICGECHTENEIKSRDPIRCRECGYRIMYKKRTKRLVVFDAR
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documenti

Documenti

Certificati
Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato