missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ POLR2K Polyclonal Antibody
Description
Immunogen sequence: DVQPPKQQPMI YICGECHTEN EIKSRDPIRC RECGYRIMYK KRTKRLV Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
RNA polymerase II (Pol II) is a multi-subunit enzyme responsible for the transcription of protein-coding genes. Transcription initiation requires recruitment of the complete transcription machinery to a promoter via solicitation by activators and chromatin remodeling factors. Pol II can coordinate 10 to 14 subunits. This complex interacts with the promoter regions of genes and a variety of elements and transcription factors. POLR2K (polymerase (RNA) II (DNA directed) polypeptide K), also known as RPB12, RPABC4, RPB7.0, hRPB7.0, hsRPB10a, RPB10alpha or ABC10-alpha, is a 58 amino acid nuclear protein belonging to the archaeal rpoP/eukaryotic RPC10 RNA polymerase subunit family. The gene encoding POLR2K is located on human chromosome 8, which consists of nearly 146 million base pairs, houses more than 800 genes and is associated with a variety of diseases and malignancies.
Specifications
Specifications
| Antigen | POLR2K |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | POLR2K |
| Gene Accession No. | P53803 |
| Gene Alias | ABC10-alpha; DNA directed RNA polymerases I, II, and III 7.0 kda polypeptide; DNA-directed RNA polymerase II subunit K; DNA-directed RNA polymerases I, II, and III subunit RPABC4; hRPB7.0; hsRPB10a; MafY; mCG145114-like; metallothionein-I gene transcription activator; Mt1a; POLR2K; polymerase (RNA) II (DNA directed) polypeptide K; polymerase (RNA) II (DNA directed) polypeptide K, 7.0kDa; polymerase (RNA) II subunit K; RNA polymerase II 7.0 kDa subunit; RNA polymerase II subunit K; RNA polymerases I, II, and III subunit ABC4; RPABC4; RPB10alpha; RPB12; RPB7.0 |
| Gene Symbols | POLR2K |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?