missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POLR3GL Monoclonal Antibody (OTI5F9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA506881S
This item is not returnable.
View return policy
Description
POLR3GL Monoclonal antibody specifically detects POLR3GL in Human samples. It is validated for Immunocytochemistry, Western BlotSpecifications
| POLR3GL | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8TEA8 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 92675 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunocytochemistry, Western Blot | |
| OTI5F9 | |
| Unconjugated | |
| DTD1 | |
| 2010003I19Rik; alternative RNA polymerase III subunit 32; DNA-directed RNA polymerase III subunit G-like; DNA-directed RNA polymerase III subunit RPC7-like; flj32422; POLR3GL; polymerase (RNA) III (DNA directed) polypeptide G (32kD)-like; polymerase (RNA) III (DNA directed) polypeptide G like; polymerase (RNA) III subunit G like; RNA polymerase III 32 kDa beta subunit; RNA polymerase III subunit C7-like; RNA polymerase III subunit G like; RPC32-beta; RPC32HOM; RPC32-like protein | |
| Full length human recombinant protein of human DTD1 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction