missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ POLR3GL Polyclonal Antibody

Product Code. 15733491 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15733491 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15733491 Supplier Invitrogen™ Supplier No. PA555570

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: VGIGKGDALP PPTLQPSPLF PPLEFRPVPL PSGEEGEYVL ALKQELRGAM RQLPYFIRPA VPKRDVERYS D Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.

DNA-dependent RNA polymerase catalyzes the transcription of DNA into RNA using the four ribonucleoside triphosphates as substrates. Specific peripheric component of RNA polymerase III which synthesizes small RNAs, such as 5S rRNA and tRNAs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POLR3GL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene POLR3GL
Gene Accession No. Q9BT43
Gene Alias 2010003I19Rik; alternative RNA polymerase III subunit 32; DNA-directed RNA polymerase III subunit G-like; DNA-directed RNA polymerase III subunit RPC7-like; flj32422; POLR3GL; polymerase (RNA) III (DNA directed) polypeptide G (32kD)-like; polymerase (RNA) III (DNA directed) polypeptide G like; polymerase (RNA) III subunit G like; RNA polymerase III 32 kDa beta subunit; RNA polymerase III subunit C7-like; RNA polymerase III subunit G like; RPC32-beta; RPC32HOM; RPC32-like protein
Gene Symbols POLR3GL
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human POLR3GL. Recombinant protein control fragment (Product #RP-94161).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84265
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.