missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POLR3H Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 280.00 - € 624.00
Specifications
| Antigen | POLR3H |
|---|---|
| Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18499211
|
Novus Biologicals
NBP2-13787-25ul |
25ul |
€ 280.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18056539
|
Novus Biologicals
NBP2-13787 |
0.1 mL |
€ 624.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
POLR3H Polyclonal specifically detects POLR3H in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| POLR3H | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| DNA-directed RNA polymerase III subunit 22.9 kDa polypeptide, DNA-directed RNA polymerase III subunit H, DNA-directed RNA polymerase III subunit RPC8, KIAA1665MGC29654, MGC111097, polymerase (RNA) III (DNA directed) polypeptide H (22.9kD), RNA nucleotidyltransferase (DNA-directed), RNA polymerase III subunit 22.9 kDa subunit, RNA polymerase III subunit C8, RNA polymerase III subunit RPC8, RPC8RPC22.9 | |
| POLR3H | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.4 ug/ml, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| Polyclonal | |
| Rabbit | |
| DNA replication Transcription Translation and Splicing | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 171568 | |
| This antibody was developed against a recombinant protein corresponding to the amino acids: GFFDDILIPPESLQQPAKFDEAEQVWVWEYETEEGAHDLYMDTGEEIRFRVVDESFVDTSPTGPSSADATTSSEELPKKEAPYTLVG | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title