missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POLR3H Monoclonal Antibody (OTI5A12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809442
This item is not returnable.
View return policy
Description
RPC8 is DNA-dependent RNA polymerase catalyzes the transcription of DNA into RNA using the four ribonucleoside triphosphates as substrates. Specific peripheric component of RNA polymerase III which synthesizes small RNAs, such as 5S rRNA and tRNAs. It is plays a key role in sensing and limiting infection by intracellular bacteria and DNA viruses. It is acting as nuclear and cytosolic DNA sensor involved in innate immune response. It can sense non-self dsDNA that serves as template for transcription into dsRNA. The non-self RNA polymerase III transcripts, such as Epstein-Barr virus-encoded RNAs (EBERs) induce type I interferon and NF-Kappa-B through the RIG-I pathway.Specifications
| POLR3H | |
| Monoclonal | |
| Unconjugated | |
| POLR3H | |
| RPC22.9, RPC8 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 171568 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI5A12 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9Y535 | |
| POLR3H | |
| Full length human recombit protein of human POLR3H produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction