missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POLR3H Monoclonal Antibody (OTI5A12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809442S
This item is not returnable.
View return policy
Description
POLR3H Monoclonal antibody specifically detects POLR3H in Human samples. It is validated for Western BlotSpecifications
| POLR3H | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8WXK3 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 79754 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Western Blot | |
| OTI5A12 | |
| Unconjugated | |
| ASB13 | |
| 5031409G22Rik; DNA-directed RNA polymerase III subunit 22.9 kDa polypeptide; DNA-directed RNA polymerase III subunit H; DNA-directed RNA polymerase III subunit RPC8; KIAA1665; POLR3H; polymerase (RNA) III (DNA directed) polypeptide H; polymerase (RNA) III (DNA directed) polypeptide H (22.9kD); polymerase (RNA) III subunit H; RNA nucleotidyltransferase (DNA-directed); RNA polymerase III subunit 22.9 kDa subunit; RNA polymerase III subunit C7; RNA polymerase III subunit C8; RNA polymerase III subunit H; RPC22.9; RPC8 | |
| Human recombinant protein fragment corresponding to amino acids 76-278 of human ASB13 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction