Learn More
Invitrogen™ POLRMT Recombinant Rabbit Monoclonal Antibody (HL3068)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556803
This item is not returnable.
View return policy
Description
POLRMT Recombinant Monoclonal Antibody for Western Blot
This gene encodes a mitochondrial DNA-directed RNA polymerase. The gene product is responsible for mitochondrial gene expression as well as for providing RNA primers for initiation of replication of the mitochondrial genome. Although this polypeptide has the same function as the three nuclear DNA-directed RNA polymerases, it is more closely related to RNA polymerases of phage and mitochondrial polymerases of lower eukaryotes.
Specifications
| POLRMT | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| O00411 | |
| POLRMT | |
| Recombinant fragment of human mtRNA polymerase. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| HL3068 | |
| Unconjugated | |
| POLRMT | |
| 1110018N15Rik; 4932416K13; APOLMT; DNA-directed RNA polymerase, mitochondrial; h-mtRPOL; LOW QUALITY PROTEIN: DNA-directed RNA polymerase, mitochondrial; mitochondrial DNA-directed RNA polymerase; MTRNAP; MTRPOL; POLRMT; polymerase (RNA) mitochondrial (DNA directed); RNA polymerase mitochondrial | |
| Rabbit | |
| Protein A | |
| RUO | |
| 5442 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |