Learn More
Invitrogen™ POLRMT Recombinant Rabbit Monoclonal Antibody (HL3068)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA556803
Product Code. 30275207
  • € 500.00 / 100µL
Description
POLRMT Recombinant Monoclonal Antibody for Western Blot
This gene encodes a mitochondrial DNA-directed RNA polymerase. The gene product is responsible for mitochondrial gene expression as well as for providing RNA primers for initiation of replication of the mitochondrial genome. Although this polypeptide has the same function as the three nuclear DNA-directed RNA polymerases, it is more closely related to RNA polymerases of phage and mitochondrial polymerases of lower eukaryotes.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
POLRMT
Recombinant Monoclonal
1 mg/mL
PBS with no preservative
O00411
POLRMT
Recombinant fragment of human mtRNA polymerase.
100 μL
Primary
Human
Antibody
IgG
Western Blot
HL3068
Unconjugated
POLRMT
1110018N15Rik; 4932416K13; APOLMT; DNA-directed RNA polymerase, mitochondrial; h-mtRPOL; LOW QUALITY PROTEIN: DNA-directed RNA polymerase, mitochondrial; mitochondrial DNA-directed RNA polymerase; MTRNAP; MTRPOL; POLRMT; polymerase (RNA) mitochondrial (DNA directed); RNA polymerase mitochondrial
Rabbit
Protein A
RUO
5442
-20°C, Avoid Freeze/Thaw Cycles