missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poly-Ubiquitin Antibody (Genentech patent anti-Polyubiquitin), mFluor Violet 500 SE, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP3-27986MFV500
This item is not returnable.
View return policy
Description
Poly-Ubiquitin Monoclonal antibody specifically detects Poly-Ubiquitin in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Poly-Ubiquitin | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| CEP80, HEL112, ribosomal protein S27a, RPS27A, S27A, UBA80, UBC, UBCEP1, UBCEP80 | |
| Polyubiquitin | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Functional Assay | |
| Genentech patent anti-Polyubiquitin | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| 6233 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction