Learn More
Masterflex™ Polycarbonate Male Luer Adapter
Brand:  Masterflex™ 45504-82
Additional Details : Weight : 2.50000kg
 View more versions of this product
Product Code. 15266464
  • € 16.05 / Pack of 25
Estimated Shipment
23-09-2020
Add to basket
Specifications
Specifications
Polycarbonate
Luer Adapter
Fits male luer to 1/4-28 thread
25 Pack