Learn More
Ekastu Safety™ K1 Plus Spare Visor
Description
- Made of 1.0mm high quality polycarbonate
- Visor with special comfort bends
Specifications
Specifications
| Thickness (Metric) | 1.0 mm |
| Lens Coating | Scratch-resistant |
| Lens Tint | Clear |
| Product Type | Replacement visor |
| For Use With (Equipment) | K1 Plus Faceshield |
| Lens Material | Polycarbonate |