missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polycystin-1/PKD1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 216.00 - € 518.00
Specifications
| Antigen | Polycystin-1/PKD1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18693550
|
Novus Biologicals
NBP2-93178-0.02ml |
0.02 mL |
€ 216.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18627820
|
Novus Biologicals
NBP2-93178-0.1ml |
0.1 mL |
€ 518.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Polycystin-1/PKD1 Polyclonal antibody specifically detects Polycystin-1/PKD1 in Human samples. It is validated for Western BlotSpecifications
| Polycystin-1/PKD1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Autosomal dominant polycystic kidney disease 1 protein, PBP, Pc-1, polycystic kidney disease 1 (autosomal dominant), polycystic kidney disease-associated protein, polycystin 1, polycystin-1, transient receptor potential cation channel, subfamily P, member 1, TRPP1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 3700-3860 of human PKD1 (NP_001009944.2). RLQSAIKQELHSRAFLAITRSEELWPWMAHVLLPYVHGNQSSPELGPPRLRQVRLQEALYPDPPGPRVHTCSAAGGFSTSDYDVGWESPHNGSGTWAYSAPDLLGAWSWGSCAVYDSGGYVQELGLSLEESRDRLRFLQLHNWLDNRSRAVFLELTRYSPA | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 5310 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto