missing translation for 'onlineSavingsMsg'
Learn More

Polydatin, MedChemExpress

Product Code. 16498986
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16498986 100 mg 100mg
16492346 500 mg 500mg
16481486 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16498986 Supplier Medchem Express Supplier No. HYN0120A
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Polydatin (Piceid), extracted from the roots of Polygonum cuspidatum Sieb, a widely used traditional Chinese remedies, possesses anti-inflammatory activity in several experimental models. Polydatin (Piceid) inhibits G6PD and induces oxidative and ER stresses.

Polydatin (Piceid), extracted from the roots of Polygonum cuspidatum Sieb, a widely used traditional Chinese remedies, possesses anti-inflammatory activity in several experimental models. Polydatin (Piceid) inhibits G6PD and induces oxidative and ER stresses.

Specifications

Chemical Name or Material Polydatin
CAS 27208-80-6
Color White
Purity Grade Notes Research
Molecular Formula C20H22O8
Quantity 100 mg
Synonym Piceid
Solubility Information DMSO : ≥ 29 mg/mL (74.29 mM) ∣H2O : 0.1 mg/mL (0.26 mM; Need ultrasonic)
SMILES O[C@H]([C@H]([C@@H]([C@@H](CO)O1)O)O)[C@@H]1OC2=CC(/C=C/C3=CC=C(O)C=C3)=CC(O)=C2.[(E)]
Molecular Weight (g/mol) 390.38
Formula Weight 390.38
Percent Purity 98.55%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.