Learn More
Brady™ Polyester Mandatory Signs
Product Code. 10265174
Change view
Click to view available options
Displayed Text:
Hands Protection
Safety Suit
Diameter (Metric):
100 mm
200 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Displayed Text Diameter (Metric) unitSize
10265174 Hands Protection 200 mm Each
10795695 Safety Suit 100 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10265174 Supplier Brady™ Supplier No. 250111
Only null left
Specifications
Alternate Part Number PIC 255-DIA 200-B7541 (BRANDED)
Material Polyester
Quantity 1 Ea.
Adhesive Type Polyethylene, Polyester
Diameter (Metric) 200 mm
Mounting Type Self-adhesive
For Use With (Application) Outdoors Use
Max. Temperature (Metric) 120°C
Displayed Text Hands Protection
Product Type Mandatory Sign
Color White on Blue