Learn More
Masterflex™ Polyethylene Elbow Pipe Adapter
Designed for quick and easy assembly without tools
€ 25.25 - € 30.40
Specifications
Product Type | Barbed Fitting |
---|---|
Description | Barb x NPT(M) Connection |
Autoclavable | Not Autoclavable |
Material | HDPE |
Packaging Quantity | 10 Fittings |
Product Code | Brand | Tubing Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Tubing Size | Price | Quantity & Availability | |||||
11765189
|
Masterflex™
30623-12 |
3/8 in. |
€ 25.25
Pack of 10 |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||||
11745549
|
Masterflex™
30623-08 |
5/8 in. |
€ 30.40
Pack of 10 |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||||
Description
- Ideal for low-pressure applications
- Chemical resistant
- Available in a variety of sizes
- Economical barbed fittings easily connect fluid paths without the use of tools
- Just slip your flexible tubing or hose onto the barb to create a secure seal
Specifications
Barbed Fitting | |
Not Autoclavable | |
10 Fittings | |
Ethylene Oxide | |
Translucent |
Barb x NPT(M) Connection | |
HDPE | |
1/2 in. | |
Elbow-shaped |