Learn More
Macherey-Nagel™ Surface Protection Paper
Provides excellent absorption for spilled liquids. Macherey Nagel™ Surface Protection Paper is filter paper coated on one side with polyethylene.
Brand: Macherey-Nagel™ 112000.1
Additional Details : Weight : 2.34000kg
Description
- For covering laboratory workbenches and tops
- Good for chemical storerooms and cupboards
- Especially suited for isotope and bacteriological labs
Specifications
Polyethylene | |
Filter Paper | |
48 cm |
50 Pack | |
48 x 60cm | |
60 cm |