Learn More
Macherey-Nagel™ Surface Protection Paper
Provides excellent absorption for spilled liquids. Macherey Nagel™ Surface Protection Paper is filter paper coated on one side with polyethylene.
Brand:  Macherey-Nagel™ 112000.1
Additional Details : Weight : 2.34000kg
Product Code. 10782511
  • € 201.00 / Pack of 50
In Stock
to see stock.
Add to basket
Description
Description
  • For covering laboratory workbenches and tops
  • Good for chemical storerooms and cupboards
  • Especially suited for isotope and bacteriological labs
Specifications
Specifications
Polyethylene
Filter Paper
48 cm
50 Pack
48 x 60cm
60 cm