missing translation for 'onlineSavingsMsg'
Learn More

B Braun™ Polyethylene Extrufix Garbage Bags

Product Code. 16497943
Click to view available options
Capacity (Metric):
15 L
30 L
Unit Size:
1 roll
Each
This item is not returnable. View return policy

Product Code. 16497943

Brand: B Braun™ 6201156

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Use B Braun™ Polyethylene Extrufix Waste Bags to dispose of garbage inside or outside the work facility.

  • Polyethylene
  • Gray

Specifications

Capacity (Metric) 15 L
Color Gray
Disposable Single-use
Material Polyethylene
Product Type Garbage Bag
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.