missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyethylene Glycol 400 Grade, Extra Pure, SLR, Fisher Chemical™
Click to view available options
Quantity:
500 mL
25 L
Packaging:
Amber glass bottle
Plastic container
Description
Polyethylene Glycol 400 Grade, Extra Pure, SLR
Specifications
pH | 5.0 to 7.0 |
Physical Form | Liquid |
Boiling Point | 200°C |
Melting Point | 4°C |
Quantity | 500 mL |
Formula Weight | 400g/mol |
Packaging | Amber glass bottle |
Vapor Pressure | <0.01mmHg at 20°C |
Viscosity | 110 to 125 mPaS at 20°C |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'