missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene Glycol 400 Grade, Extra Pure, SLR, Fisher Chemical™

Catalog Number 10744381
Click to view available options
Quantity:
500 mL
25 L
Packaging:
Amber glass bottle
Plastic container
This item is not returnable. View return policy
25322-68-3
(C2H4O)n
62.07
MFCD01779601
LYCAIKOWRPUZTN-UHFFFAOYSA-N
ethane-1,2-diol
[H]OCCO
This item is not returnable. View return policy
25322-68-3
(C2H4O)n
62.07
MFCD01779601
LYCAIKOWRPUZTN-UHFFFAOYSA-N
ethane-1,2-diol
[H]OCCO

Polyethylene Glycol 400 Grade, Extra Pure, SLR

TRUSTED_SUSTAINABILITY
pH 5.0 to 7.0
Physical Form Liquid
Boiling Point 200°C
Melting Point 4°C
Quantity 500 mL
Formula Weight 400g/mol
Packaging Amber glass bottle
Vapor Pressure <0.01mmHg at 20°C
Viscosity 110 to 125 mPaS at 20°C
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'