missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene glycol dimethanesulfonate, M.W. 8,000, Thermo Scientific™

Catalog Number p-7080121
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
52410-74-9
C4H10O6S2
218.238
MFCD03100947
QSQFARNGNIZGAW-UHFFFAOYSA-N
ethylene dimethanesulfonate, dmse, 1,2-bis mesyloxy ethane, ethylene methanesulfonate, ethylene glycol dimesylate, ethane dimethane sulphonate, 1,2-ethanediyl dimethanesulfonate, dimethylene dimethanesulfonate, ethylene bis methanesulfonate, ethane dimethanesulphonate
20796
2-methylsulfonyloxyethyl methanesulfonate
CS(=O)(=O)OCCOS(=O)(=O)C
This item is not returnable. View return policy
52410-74-9
C4H10O6S2
218.238
MFCD03100947
QSQFARNGNIZGAW-UHFFFAOYSA-N
ethylene dimethanesulfonate, dmse, 1,2-bis mesyloxy ethane, ethylene methanesulfonate, ethylene glycol dimesylate, ethane dimethane sulphonate, 1,2-ethanediyl dimethanesulfonate, dimethylene dimethanesulfonate, ethylene bis methanesulfonate, ethane dimethanesulphonate
20796
2-methylsulfonyloxyethyl methanesulfonate
CS(=O)(=O)OCCOS(=O)(=O)C

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color White
Physical Form Granular Solid
Quantity 250 mg
Chemical Name or Material Polyethylene glycol dimethanesulfonate, M.W. 8,000
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'