missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyethylene glycol (methoxy group) Rabbit, Clone: RM105 (Biotin), Abnova™
Rabbit Monoclonal Antibody
Brand: Abnova MAB14965.50ug
This item is not returnable.
View return policy
Beschreibung
Rabbit monoclonal antibody raised against Polyethylene glycol (methoxy group).
- High specificity
- High reactivity
Spezifikation
| Polyethylene glycol (methoxy group) | |
| Monoclonal | |
| Biotin | |
| ELISA (0.02-0.5 ug/mL) Flow Cytometry Immunocytochemistry Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-2 ug/mL) Immunoprecipitation Western Blotting (0.1-1 ug/mL) The optimal working dilution should be determ | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot | |
| RM105 | |
| Rabbit monoclonal antibody raised against Polyethylene glycol (methoxy group). | |
| In PBS (50% glycerol, 1% BSA, 0.09% sodium azide) | |
| Polyethylene glycol (methoxy group) conjugated with KLH. | |
| 50 μg | |
| Primary | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only (RUO)
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur