Learn More
Polyethylene oxide, M.W. >5,000,000
Description
It is used as a thickening agent and gel binder in the pharmaceutical industry. It is used as a analytical reagents, substrates, flocculants, dispersants, lubricants, emulsifiers, moisturizers, food additives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| pH | 5 to 7 |
| Melting Point | ∼175°C |
| Solubility Information | Soluble in water. |
| Quantity | 500 g |
| Formula Weight | >5,000,000 |
| Odor | Odorless |
| Chemical Name or Material | Polyethylene oxide |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.