Learn More
Hirschmann™ Polyethylene Poly Stopper
Product Code. 30090421
Click to view available options
Standard Taper Joint:
NS 10/19
NS 12/21
NS 14/23
NS 45/40
NS 7/16
Unit Size:
Each
Pack of 10
Product Code. 30090421
Brand: Hirschmann™ 8900252
Only null left
Add to basket
Edge
Add to basket
For the safe and secure closure of flasks, bottles, tubing and other laboratory apparatus.
  • PE
  • Available to fit a range of sizes
Specifications
Quantity 1 Ea.
Standard Taper Joint NS 7/16
Material Polyethylene
Product Type Polyethylene Stopper
Shape Hexagonal Grip