missing translation for 'onlineSavingsMsg'
Learn More

Ark Plas Products™ Polyethylene Tubing

An economical choice for nonconductive tubing in the laboratory, providing good resistance to chemicals.

Brand:  Ark Plas Products™ TEX75-TE0

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11753884

  • € 148.13 / Each


Estimated Shipment
09-04-2021
Add to basket

Description

Description

  • Structure helps maintain flexibility and durability, even at low temperature
  • Broad resistance to chemicals, can be used as a fluid line for liquids or gases
  • Manufactured from resin, natural in color
  • Extruded using state of the art inline process monitoring equipment
  • Shipped bulk on cardboard spools
Specifications

Specifications

Natural
15.87 mm
19.05 mm
1.57 mm
Autoclavable
Polyethylene
100 m
1 Pack
Videos
SDS
Documents