Learn More
Scancell Folien Vertriebs™ Polyethylene Ziplock Bag
Product Code. 10460233
Click to view available options
Dimensions (L x W):
150 x 220 x 0.05mm
400 x 500 x 0.05mm
Unit Size:
Pack of 100
Pack of 1000
Product Code. 10460233
Brand: Scancell Folien Vertriebs™ 11001522
Only null left
Add to basket
Edge
Add to basket
Specifications
Color Clear
Material Polyethylene
Thickness (Metric) 0.050 mm
Quantity 100 Pack
Dimensions (L x W) 150 x 220 x 0.05mm
Product Type Ziplock Bag