Learn More
Polygalacturonic acid, M.W. 25,000-50,000
Description
Polygalacturonic acid is reagent which sequesters metal ions and produces gels that can be used for the production of biomolecules. It is used for drug release or to immobilize cells and enzymes . Polygalacturonic acid may be used to identify, differentiate and characterized plant cell wall degrading polygalacturonase(s) from fungi.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Solubility Information | Soluble in water. |
| Quantity | 500 g |
| Formula Weight | 25 to 50 kd |
| Optical Rotation | +250° (c=1 in 0.1N NaOH) |
| Sensitivity | Hygroscopic |
| Chemical Name or Material | Polygalacturonic acid |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.