Specifications
| Particle Size | 40 to 63 μm |
| Pore Size | 100 Å |
| Stationary Phase | POLYGOPREP 100 to 50 |
| Technique | Liquid Chromatography |
| pH | 2 to 8 |
| Quantity | 1 kg |
| Sorbent | POLYGOPREP |
| Surface Area (Metric) | 280 m2/g |
| Product Type | Polygoprep |
| Material | Silica |
| Show More |