Learn More
Macherey-Nagel™ Polygoprep™ 1000 to 30 C18
Shop All Macherey-Nagel Products
Click to view available options
Quantity:
0.1 kg
1 kg
Particle Size:
25 to 40 μm
40 to 63 μm
Unit Size:
1000g
100g
Specifications
Specifications
| Particle Size | 25 to 40 μm |
| Pore Size | 1000 Å |
| Stationary Phase | POLYGOPREP 1000 to 30 |
| pH | 2 to 8 |
| Quantity | 0.1 kg |
| Sorbent | POLYGOPREP C18 |
| Type | Polygoprep |
| For Use With (Application) | Liquid Chromatography |